Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pig Trypsin (from Pancreas)
Pig Trypsin (from Pancreas)
Pig Trypsin (from Pancreas)
650279-1SET
:
Merck Millipore (Calbiochem)
:
Pig Trypsin (from Pancreas)
650279-1SET
:
Merck Millipore (Calbiochem)
:
Pk:
1 SET
CAS No.:
9002-07-7
Source:
Pancreas
Species:
Pig
Storage conditions:
Refrigerator
MDL No.:
MFCD00082094
Enzyme name:
Trypsin