Bovine trypsin (from pancreas)
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Specifications
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Bovine
- Storage conditions:Refrigerator
- MDL No.:MFCD00082094
- Enzyme name:Trypsin
Specifications
Frequently Bought Together