Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Starch 1% in aqueous solution

Glass bottle, 500 mL
Catalog # ROTHT133.1
Marketsource
Supplier:   Roth Carl
Starch 1% in aqueous solution
Catalog # ROTHT133.1
Supplier:  Roth Carl
Marketsource
CAS Number:  9005-25-8
Starch 1% in aqueous solution

Glass bottle, 500 mL

CAS Number:  9005-25-8
Price
245,69 kr
Log in for availability
1 * 500 mL
Product Details & Documents
  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-679-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications