Order Entry
Denmark
ContactUsLinkComponent
Riboflavine (vitamin B2) E101 (i), SAFC®
Riboflavine (vitamin B2) E101 (i), SAFC®
Catalog # 5.00257.2500
CAS Number:  
Riboflavine (vitamin B2) E101 (i), SAFC®
Catalog # 5.00257.2500
CAS Number:  

Specifications

  • Pack type:
    Metal can
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

About this item

 :
 :376.37 g/mol
Merck Index:12,08367
Melting Pt:290 °C (dec.)
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:201-507-1
CAS Number:83-88-5
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022