Order Entry
Denmark
ContactUsLinkComponent
Potassium chromate 5% in aqueous solution Reag. Ph. Eur. 1069201
Potassium chromate 5% in aqueous solution Reag. Ph. Eur. 1069201
Catalog # 87899.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium chromate 5% in aqueous solution Reag. Ph. Eur. 1069201
Catalog # 87899.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :194.19 g/mol
EINECS:232-140-5
UN:3082
ADR:9,III
 :GHS08
 :GHS09
CAS Number:7789-00-6
Formula:K₂CrO₄
MDL Number:MFCD00011368