


Lead(II) perchlorate trihydrate 97.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13453-62-8
:
Formula:Cl₂O₈Pb·3H₂O
MDL Number:MFCD00149817
:460.15 g/mol
Melting Pt:100 °C (dec.)
Density:2.60
EINECS:237-125-7
UN:1470
ADR:5.1,II
:GHS03
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together