



Lead(II) perchlorate trihydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13453-62-8
:
Formula:Cl₂O₈Pb·3H₂O
MDL Number:MFCD00149817
:460.15 g/mol
Melting Pt:100 °C (dec.)
Density:2.6 g/cm³
Storage Temperature:Ambient
EINECS:237-125-7
UN:1470
ADR:5.1,II
:GHS07
:GHS08
:GHS09
:GHS03
Specification Test Results
- Appearance:White to light beige Adhering crystals
- Titration Complexometric:≥98.5 %
- pH:3 to 5 (5 % solution)
- Chloride (Cl):≤100 ppm
- Iron (Fe):≤10 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together