Rhodamine B, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 1389.0100
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Sensitivity as Indicator:passes test
- Solubility:passes test
Specification Test Results
About this item
Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
:
:479.02 g/mol
Merck Index:13,08266
Melting Pt:199…201 °C
Density:1.31 g/cm³ (20 °C)
EINECS:201-383-9
:GHS05
Storage Temperature:Ambient
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931