Bromophenol blue
Glass bottle, 250 mL
Catalog # CHRO3L-105
Marketsource
Supplier: CHROMA
Jump to:
Product Details & Documents
Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
- CAS Number:115-39-9
- Formula:C₁₉H₁₀Br₄O₅S
- MDL Number:MFCD00005875
- Molecular Weight:669.97 g/mol
- Merck Index:13,01429
- Melting Pt:273 °C
- Density:1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:204-086-2
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



