
Bromocresol green sodium salt, water soluble indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67763-24-0
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898
Melting Pt:230 °C (dec.)
:720 g/mol
EINECS:263-657-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark brown to green-black Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Visual transition interval:from pH 3.8 (yellow) to pH 5.4 (blue)
- Clarity of solution:(0.1% in water) Clear
Specification Test Results
Frequently Bought Together