Order Entry
Denmark
ContactUsLinkComponent
APF ~5mM ≥98% (by HPLC)
APF ~5mM ≥98% (by HPLC)
  ENZOALX620075M001
 :  
APF ~5mM ≥98% (by HPLC)
  ENZOALX620075M001
 :  

 

  • Pk:
    1 mg
  • Seq:
    2070

 

 

  • Concentration:
    ~5 mM
  • Formulation:
    Dissolved in 0.47 ml dimethyl formamide
  • Purity:
    ≥98% (HPLC)

 

 

 :
 :3,III
 :423.4 g/mol
 :200-679-5
 :2265
 :GHS07
 :GHS08
 :Refrigerator
 :359010-70-1
 :C₂₆H₁₇NO₅