Order Entry
Denmark
ContactUsLinkComponent
Galantamine hydrobromide ≥98% (by HPLC)

Specifications

  • Pk:
    50 mg
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Galanthamine HBr, (4aS,6R,8aS)-3-Methoxy-11-methyl-5,6,9,10,11,12-hexahydro-4aH-benzo[2,3]benzofuro[4,3-cd]azepin-6-ol hydrobromide
 :
 :368.27 g/mol
Storage Temperature:Freezer
CAS Number:1953-04-4
Formula:C₁₇H₂₁NO₃·HBr
MDL Number:MFCD00067672