EMPO ≥95% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Synonyms: 2-Ethoxycarbonyl-2-methyl-3,4-dihydro-2H-pyrrole-1-oxide
CAS Number:61856-99-3
:
Formula:C₈H₁₃NO₃
Molecular Weight:171.2 g/mol
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity (NMR,HPLC):≥95%
Specification Test Results
Frequently Bought Together