Order Entry
Denmark
ContactUsLinkComponent
Thiomersal
Thiomersal
Catalog # ENZOALX400013G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Thiomersal
Catalog # ENZOALX400013G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :404.82 g/mol
Merck Index:12,09451
Melting Pt:232…233 °C
Flash Pt:250 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-210-4
UN:2025
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:54-64-8
Formula:2-(C₂H₅HgS)C₆H₄CO₂Na
MDL Number:MFCD00013062