Order Entry
Denmark
ContactUsLinkComponent
Rufloxacin ≥98%
Rufloxacin ≥98%
Catalog # ENZOALX380300M250
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Rufloxacin ≥98%
Catalog # ENZOALX380300M250
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

 :
 :363.4 · 36.5 g/mol
CAS Number:106017-08-7
Formula:C₁₇H₁₈FN₃O₃S·HCl