Nomilin ≥98% (by HPLC)
Catalog #
Product Details & Documents
Carciogenesis inhibitor
Induces phase II detoxifying enzymes and inhibits chemically induced carcinogenesis. Exhibits immunomodulatory activity and also inhibits HIV-1 replication. Does not possess any inherent antioxidant capacity.
Isolated from grapefruit seed and citrus juice.
Synonyms: (1S,3aS,4aR,4bR,6aR,11S,11aR,11bR,13aS)-1-(Furan-3-yl)-4b,7,7,11a,13a-pentamethyl-3,5,9-trioxohexadecahydrooxepino[4',3':3,4]benzo[1,2-f]oxireno[2,3-d]isochromen-11-yl acetate
- CAS Number:1063-77-0
- Formula:C₂₈H₃₄O₉
- MDL Number:MFCD00075979
- Molecular Weight:514.5600 g/mol



