Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fumitremorgin C
Fumitremorgin C ≥96% (by HPLC)
Fumitremorgin C ≥96% (by HPLC)
Fumitremorgin C ≥96% (by HPLC)
ENZOALX350127C250
:
ENZO LIFE SCIENCES
New Product
:
Fumitremorgin C ≥96% (by HPLC)
ENZOALX350127C250
:
ENZO LIFE SCIENCES
New Product
:
Pk:
250 µG
Seq:
1370
Environmentally Preferable:
:
:
379.5 g/mol
:
118974-02-0
:
C₂₂H₂₅N₃O₃