NPC-15437 dihydrochloride ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:136449-85-9
:
Frequently Bought Together
Screw caps for laboratory/media bottles
Supplier: SciLabware
VWR®, Centrifuge Tubes, Light-Sensitive
Supplier: VWR Collection
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR®, Cryovials
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Cell culture dishes, Nunc™
Supplier: Thermo Fisher Scientific
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
BOTTLE 60ML PS NA, H55 Ø40 1 * 500 items
Supplier: Corning
VWR®, Cell Culture Flasks, Non Treated or Treated
Supplier: VWR Collection
96 MicroWell™ plates, Nunclon™∆
Supplier: Thermo Fisher Scientific
Cup, large with handle
Supplier: LAMAPLAST
VWR®, Microcentrifuge Tubes with Socket Screw Caps
Supplier: VWR Collection
VWR® Cryoboxes, Double Size, 133x257 mm
Supplier: VWR Collection