bisBenzimide H 33258 trihydrochloride (Hoechst 33258) ≥95% (by HPLC), Ultrapure dye
Katalog # ENZOENZ52402
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:533.88 g/mol
Storage Temperature:Freezer
CAS Number:23491-45-4
Formula:C₂₅H₂₄N₆O·3HCl
MDL Number:MFCD00012679