Order Entry
Denmark
ContactUsLinkComponent
Lysophosphatidic acid ≥98% (by TLC)
Lysophosphatidic acid ≥98% (by TLC)
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Lysophosphatidic acid ≥98% (by TLC)
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Purity:
    ≥98% (TLC)
  • Appearance:
    Waxy solid

Specification Test Results

Frequently Bought Together