C2 Ceramide ≥98% (by TLC)
Catalog #
Product Details & Documents
Cell-permeable ceramide analog
N-Acetylsphingosine is a biologically active, cell-permeable ceramide analog. It inhibits cell proliferation and induces monocytic differentiation of HL-60 cells and induces apoptosis. It stimulates protein phosphatase 2A4, activates MAP kinase and SAP kinase and induces PKCδ and ε translocation. Physiological levels of C2 ceramide, produced by the action of a PAF:sphingosine CoA-independent transacetylase, have been detected in HL-60 cells.
- CAS Number:3102-57-6
- Formula:C₂₀H₃₉NO₃
- MDL Number:MFCD00153903
- Molecular Weight:341.536 g/mol



