Order Entry
Denmark
ContactUsLinkComponent
Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)
Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)
Catalog # ENZOBMLGR3000005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Actinomycin D (from Streptomyces parvulus) ≥98% (by TLC)
Catalog # ENZOBMLGR3000005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :1255.5 g/mol
Storage Temperature:Freezer
UN:3462
ADR:6.1,II
 :GHS06
CAS Number:50-76-0
Formula:C₆₂H₈₆N₁₂O₁₆
MDL Number:MFCD00005033