Order Entry
Denmark
Orders LinkContactUsLinkComponent
Vacu-vials® Snapper
Vacu-vials® Snapper
Vacu-vials snapper
Catalog # CHETA-0214
Supplier:   CHEMetrics
Vacu-vials® Snapper
Catalog # CHETA-0214
Supplier:  CHEMetrics
Vacu-vials® Snapper
Vacu-vials snapper
Price
827,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
This reusable vial snapper permits beverage manufacturers to conveniently monitor disinfectant and sterilisation residual levels directly inside a bottle or carton taken off the production line. Since the measurement is obtained in a single container, sample agitation is minimised leading to more accurate measurements.
Specifications
  • Description:
    Vacu-vials snapper