Order Entry
Denmark
Orders LinkContactUsLinkComponent
Boss Head
Boss Head
Cross sleeve (2 clamps are required to attach the SilentCrusher M and the vessel holding bracket)
Catalog # 431-0043
Boss Head
Catalog # 431-0043
Boss Head
Boss Head
Cross sleeve (2 clamps are required to attach the SilentCrusher M and the vessel holding bracket)
Price
332,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Boss head made of anodised aluminum, used to fix retort clamps to stand rods for perfect vertical alignment.
Suitable for stand rods with up to 13,1 mm maximum diameter.
Specifications
  • Description:
    Cross sleeve (2 clamps are required to attach the SilentCrusher M and the vessel holding bracket)