Cholesterol
Glass bottle for solids, 100 g
Catalog # SERA17101.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
- CAS Number:57-88-5
- Formula:C₂₇H₄₆O
- MDL Number:MFCD00003646
- Molecular Weight:386.66 g/mol
- Merck Index:13,02221
- Boiling Pt:360 °C (1013 hPa)
- Melting Pt:147…150 °C
- Density:1.07 g/cm³ (20 °C)
- Storage Temperature:Freezer
- EINECS:200-353-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



