Multirisks Cabinents, F30
Supplier: JINVALL INREDNINGAR
Certificates
About this item
The cabinets are divided into several different sections. When we want to store different substances that may not be stored together.
- Self-closing doors that can be set up and close automatically when the temperature rises to 50 °C
- Ventilation openings: Ø 100 mm
- Doors with three-point locking and cylinder lock
For storage of all chemicals that are often used in e.g. laboratory and industries and which should be kept separately. The inside is divided by a wall which makes it possible to use the cabinets for several different areas. The cupboard is equipped, depending on the model, with troughs, shelves or perforated bottom plate. Can also be furnished with PVC inserts when storing corrosive substances.
Certifications: Fire tested according to EN 14470-1.AN105. Approved according to FM standard 6050.
Frequently Bought Together