Order Entry
Denmark
ContactUsLinkComponent
L-Alanine ≥99%, high purity
L-Alanine ≥99%, high purity
Catalog # 0106-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
L-Alanine ≥99%, high purity
Catalog # 0106-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Arsenic:
    ≤0.00015%
  • Ash:
    ≤0.15%
  • Chloride:
    ≤0.05%
  • Heavy Metals:
    ≤0.001%
  • Sulphate:
    <0.03%
  • Iron:
    <0.003%
  • Loss on Drying:
    <0.2%
  • pH (5%, Water) @25 °C:
    5.5 - 7.0
  • Purity:
    >99%
  • Specific Rotation:
    13.7 -15.1°

Specification Test Results

About this item

 :
 :89.09 g/mol
Merck Index:13,00203
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-273-8
CAS Number:56-41-7
Formula:C₃H₇NO₂
MDL Number:MFCD00064410