L-Alanine, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:56-41-7
:
Formula:C₃H₇NO₂
MDL Number:MFCD00064410
:89.09 g/mol
Merck Index:13,00203
Boiling Pt:189 °C (1013 hPa)
Melting Pt:79 °C
Density:1.4 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-273-8
Related Information
Frequently Bought Together