Order Entry
Denmark
ContactUsLinkComponent
Stoppers, Glass, Conical Ground Joint
Stoppers, Glass, Conical Ground Joint
Catalog # 217-0767
Supplier:  DWK Life Sciences
Stoppers, Glass, Conical Ground Joint
Catalog # 217-0767
Supplier:  DWK Life Sciences
Specifications
  • NS:
    19/26
  • Type:
    Clear glass
  • Pk:
    10
Specifications
About this item
DURAN® glass stopper with conical ground joint, for reagent bottles and oxygen bottles according to Winkler.
  • Made of borosilicate 3.3 glass