Order Entry
Denmark
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate ≥98.0%, crystals USP
Disodium hydrogen phosphate heptahydrate ≥98.0%, crystals USP
Catalog # SPCMSO140-12KG
Supplier:  Spectrum Chemical
CAS Number:  
Disodium hydrogen phosphate heptahydrate ≥98.0%, crystals USP
Catalog # SPCMSO140-12KG
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly pail
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.0 - 100.5 %
  • Loss On Drying:
    43.0 - 50.0 %
  • Insoluble Matter:
    max. 0.4 %
  • Chloride (Cl):
    max. 0.06 %
  • Sulfate (SO₄):
    max. 0.2 %

Specification Test Results

About this item

 :
 :268.07 g/mol
Storage Temperature:Ambient
CAS Number:7782-85-6
Formula:HNa₂O₄P·7H₂O
MDL Number:MFCD00149180