Order Entry
Denmark
ContactUsLinkComponent
Anti-B7-H3/CD276 Rabbit Polyclonal Antibody
Anti-B7-H3/CD276 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-88966-25
Supplier:  Novus Biologicals
Anti-B7-H3/CD276 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-88966-25
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    B7-H3/CD276
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    B7-H3B7 homolog 3|CD276 antigen|B7H34Ig-B7-H3|Costimulatory molecule|CD276 molecule
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at 4 °C short term, aliquot and store at −20 °C long term, avoid freeze-thaw cycles
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids: EVQVPEDPVVALVGTDATLCCSFSPEPGFSLAQLNLIWQLTDTKQLVHSFAEGQDQGSAYANRTALFPDLLAQGNASLRLQRVRVADEGSFTCFVSIRDFG
  • Purification:
    Immunogen affinity purified
  • Pk:
    25 µl

Specifications

About this item

The B7-H3/CD276 antibody is a rabbit polyclonal antibody to B7-H3/CD276. This antibody reacts with human. The B7-H3/CD276 antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: B7-H3/CD276
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human