D-Lysine ≥95.0% (by titration analysis on dried basis)
Catalog # TCIAL0173-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity(Nonaqueous Titration):min. 95.0 %
- Specific rotation:-24.0 to -27.0 deg(C=1, 5mol/L HClaq.)
- NMR:confirm to structure
Specification Test Results
About this item
:
:146.19 g/mol
Melting Pt:218 °C (dec.)
Storage Temperature:Room Temperature
EINECS:213-091-9
CAS Number:923-27-3
Formula:C₆H₁₄N₂O₂
MDL Number:MFCD00008234