Human Recombinant SCF
ORIGTP723730
: OriGene
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Storage conditions:Store at −80 °C.
- Endotoxin content:Less than 0,01 ng per µg protein as determined by the LAL method.
- Biological activity:The ED50 is 3 - 12 ng/ml, corresponding to a specific activity of 0,8 - 3,3×10⁵ units/mg, determined by TF-1 cell proliferation induced in a dose dependent manner.
- Endotoxin-free:Yes
- Protein synonyms:FPHH|SLF|KL-1|Kitl|DFNA69|DCUA|SF|FPH2|MGF|SHEP7
- Protein/peptide name:SCF
- Purity:>98%, as determined by Coomassie stained SDS-PAGE.
- Molecular weight:18,6 kDa
- Concentration:Lot specific
This gene encodes the ligand of the tyrosine-kinase receptor encoded by the KIT locus. This ligand is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis. Two transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq, Jul 2008).