Order Entry
Denmark
ContactUsLinkComponent
Crack Set for Heavy Metal Digestion
Crack Set for Heavy Metal Digestion
  WTWA250496
 :  WTW
 :  
Crack Set for Heavy Metal Digestion
  WTWA250496
 :  WTW
 :  
  • Pk:
    1 KIT
  • Description:
    Crack set 10 for thermal digestion of heavy metals, sufficient for 100 digestions
  • Type:
    Crack-SET 10 14687
  • Contents:
    Reagent for 100 digestions in 16 mm empty cells