Order Entry
Denmark
ContactUsLinkComponent
Methylene blue solution R in ethanol 95%
Flammable
Irritant
Methylene blue solution R in ethanol 95%
Catalog # 1808-50ML
CAS Number:  
Flammable
Irritant
Methylene blue solution R in ethanol 95%
Catalog # 1808-50ML
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :319.86 g/mol
Flash Pt:13 °C
 :GHS02
 :GHS07
CAS Number:61-73-4
 :MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008