Order Entry
Denmark
ContactUsLinkComponent
Corning® Elplasia® 12K Open Well Plate
Corning® Elplasia® 12K Open Well Plate
Catalog # CORN4547
Supplier:  Corning
undefined
Corning® Elplasia® 12K Open Well Plate
Catalog # CORN4547
Supplier:  Corning

Specifications

  • No. of wells:
    12000 microcavities
  • Recommended working volume:
    20 - 30 ml
  • Sterile:
    +
  • Description:
    Corning® Elplasia® 12K open well plate
  • Pk:
    5

Specifications

About this item

Culture thousands of spheroids in a microplate footprint.

  • Ease of spheroid formation, culture, assessment, and harvest
  • Ready-to-use plate to create large quantities of uniform spheroids
  • Scaffold-free cultures
  • Culture spheroids for up to 14 or more days (cell line-dependent)
  • Highly reproducible bulk spheroid formation across microcavities
  • Common media reservoir for equivalent culture conditions for all spheroids
  • Easy access to spheroid cultures
  • Compatible with brightfield and fluorescent microscopy

Critical in drug screening and other applications such as cell therapy research, spheroids are coveted for their ability to more closely resemble the original tumor from which they were derived. The Corning Elplasia 12K plate is designed similarly to the Corning Elplasia 12K flask, but in a microplate footprint which makes it ideal for easy sampling and imaging of spheroids. The plate also features media exchange ports for ease of liquid handling.

No. of microcavities: 12000
Microcavity dimensions: 500W×600 µm D
Surface treatment: Ultra-low attachment
Material: Polystyrene

Certifications: Sterile.
Packaging: 1/pack, 5/case.