Accessories for Visible Spectrophotometer, pHotoLab® 7100
: WTW
Frequently Bought Together
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Ethanol 70% ≥70% (v/v)
: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
: VWR Chemicals
Formalin 10%, Q Path®, buffered
: VWR Chemicals
VWR®, OCT Embedding matrix for frozen sections
: VWR Chemicals
VWR® Dosatest, pH indicator strips for coloured samples
: VWR Chemicals
L(+)-Ascorbic acid
: Hach