Order Entry
Denmark
Orders LinkContactUsLinkComponent
Ammonia solution

Ammonia solution

1 L
Catalog # CMPL1004705.1000
Corrosive
Irritant
Ammonia solution
Catalog # CMPL1004705.1000
CAS Number:  1336-21-6
Ammonia solution

1 L

CAS Number:  1336-21-6
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.

Instruction for use: The solution bottle should be open for the minimum time required to dispense the solution. After use, the bottle should be tightly recapped and stored under normal laboratory conditions.

Shelf Life on Ship Date: 12 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.

Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pk:
    1 L

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.