
Octadecyl derivatized silica 15-40 μm mean particle size, average diameter 60 Å, Supelco®
Catalog # H8261-100G
Supplier: Merck
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:66369-14-0
MDL Number:MFCD00166485