Order Entry
Denmark
ContactUsLinkComponent
 
Xylitol
Catalog # EHERC17945500
Supplier:  EHRENSTORFER
CAS Number:  
Xylitol
Catalog # EHERC17945500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :152.15 g/mol
Merck Index:13,10140
Boiling Pt:495 °C (1013 hPa)
Melting Pt:94 °C
Density:1.52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-788-0
CAS Number:87-99-0
Formula:C₅H₁₂O₅
MDL Number:MFCD00064292