Bis(4-chlorophenyl)disulphide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1142-19-4
:
Formula:C₁₂H₈Cl₂S₂
MDL Number:MFCD00013642
:287.23 g/mol
Melting Pt:71…74 °C
Storage Temperature:Ambient
EINECS:214-531-2
Frequently Bought Together