Order Entry
Denmark
Orders LinkContactUsLinkComponent
Citric acid monohydrate

Citric acid monohydrate

Katalog #
Irriterende
Citric acid monohydrate
Katalog #
Leverandør:  
CAS nummer:  5949-29-1
Citric acid monohydrate
Citric acid monohydrate
CAS nummer:  5949-29-1
Product Details & Documents
Synonymer:  2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
  • CAS nummer:5949-29-1
  • Formel:C₆H₈O₇.H₂O
  • MDL nummer:MFCD00149972
  • Molecular Weight:210.14 g/mol
  • Merck Index:13,02350
  • Smeltende Pt:135…152 °C
  • Tæthed:1.552 g/cm³ (20 °C)
  • Flash Pt:173.9 °C (closed cup)
  • Opbevaringstemperatur:Ambient
  • EINECS:201-069-1