Organic reference standard, 1,4-Butanediol 1000 µg/ml in methanol, Cerilliant
CERIB-020
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:110-63-4
- Description:1,4-Butanediol 1,000 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₄H₁₀O₂
- Description:1,4-Butanediol