Order Entry
Denmark
ContactUsLinkComponent
Human Recombinant Acylphosphate Phosphohydrolase 2 (from E. coli)
Human Recombinant Acylphosphate Phosphohydrolase 2 (from E. coli)
Catalog # PRSI92-307
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant Acylphosphate Phosphohydrolase 2 (from E. coli)
Catalog # PRSI92-307
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-GST tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt, please minimise freeze - thaw cycles
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Enzyme name:
    Acylphosphate phosphohydrolase 2
  • Enzyme synonyms:
    Acylphosphatase-2, ACYP2, Acylphosphatase, muscle type isozyme, Acylphosphate phosphohydrolase 2, ACYP
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    37,43 kD
  • Sequence:
    Ser2-Tyr99
  • Formulation:
    Supplied as a 0,2 µm filtered solution of PBS,pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

ACYP2, also known as Acylphosphatase-2, Acylphosphatase, muscle type isozyme, Acylphosphate phosphohydrolase 2, is a protein which belongs to the acylphosphatase family.ACYP2 contains one acylphosphatase-like domain.

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: N-GST tag

Muscle acylphosphatase and erythrocyte acylphosphatase have been isolated.ACYP2 plays an important role in hydrolyzing the phosphoenzyme intermediate of different membrane pumps. Recombinant human ACYP2 protein, fused to GST-tag at N-terminus, was expressed in E. coli.