Order Entry
Denmark
Orders LinkContactUsLinkComponent
Recipient
Recipient
Recipient
Catalog # 765-0146
Marketsource
Recipient
Catalog # 765-0146
Marketsource
Recipient
Recipient
Recipient
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents
Recipient with opening at the top. With the help of a rubber plug with electrical connection bushings, for example, a bell can be hung up in ordinary laboratory cables.
  • Height, inv: 300 mm
  • Diameter, inner: 200 mm
Specifications
  • Description:
    Recipient
  • Cat. No.:
    765-0146