Order Entry
Denmark
ContactUsLinkComponent
Measuring glass with three outlets
Measuring glass with three outlets
Catalog # 765-0141
undefined
Measuring glass with three outlets
Catalog # 765-0141

Specifications

  • Ø:
    60 mm
  • Type:
    Glass with 3 outlets
  • Height:
    330 mm
  • Pk:
    1

Specifications

About this item

Transparent measuring glass with outlet at different levels. Used, among other things, for density determination, pressure and vacuum tests and optical tests. The outlets can be closed with the supplied plugs.

Height: 330 mm

Diam: 60 mm