Air density balls
765-0150
- Description:Kit for determination of air density
- Pk:1
Consists of a ball with check valve, vent valve and suspension hook. For use in conjunction with an electronic scale (0,001 g).
- Ball diameter: 90 mm
Consists of a ball with check valve, vent valve and suspension hook. For use in conjunction with an electronic scale (0,001 g).