Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pendulum balls

Pendulum balls

Marketsource
Pendulum balls
Marketsource
Certificates
Pendulum balls

5 Options Available Below

Product Details & Documents
Product Family Options

Product Information

  • Material
    Ø
    Length
    Availability
    Price
  • 765-0128
    Pendulum ball, steel, 25 mm
    Pendulum ball, steel, 25 mm
    Steel
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

    Specifications:
    Marketsource
    Marketsource
  • 765-0130
    Pendulum ball, steel, 25 mm
    Pendulum ball, steel, 25 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

    Specifications:
    Marketsource
    Marketsource
  • 765-0132
    Pendulum ball, steel, 25 mm
    Pendulum ball, steel, 25 mm
    Lead
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

    Specifications:
    Marketsource
    Marketsource
  • 765-0135
    Pendulum ball, wood, 60 mm
    Pendulum ball, wood, 60 mm
    Wood
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

    Specifications:
    Marketsource
    Marketsource
  • 765-0134
    Pendulum ball, steel, 60 mm
    Pendulum ball, steel, 60 mm
    Plastic
    60 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.