Order Entry
Denmark
Orders LinkContactUsLinkComponent
Daniell element
Daniell element
Daniell element
Catalog # 765-0199
Marketsource
Daniell element
Catalog # 765-0199
Marketsource
Daniell element
Daniell element
Daniell element
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Used in electrochemistry. Voltage, max 1 V. The unit is divisible and consists of glass beaker, cylindrical zinc electrode, plastic beaker, cylindrical copper electrode and crocodile clamps. Fill with copper sulphate (10%) or zinc sulphate (10%). A light-rotating motor with a propeller is suitable as a voltage indicator.
Specifications
  • Description:
    Daniell element