Order Entry
Denmark
ContactUsLinkComponent
Rubber belts and drive belts for generators
Rubber belts and drive belts for generators
Catalog # 765-0934
undefined
Rubber belts and drive belts for generators
Catalog # 765-0934
Specifications
  • Description:
    Drive belt, Norstedt, 12 V
  • Pk:
    1
Specifications
About this item
765-0325 Rubber band, Norstedt; Fits Norstedt's belt generators.
  • Dimensions: 100 × 7 cm
765-0934 Drive belt, Norstedt, 12 V; Fits Norstedt's belt generators with 12 V motor
765-1354 Drive belt, Norstedt, 220 V; Fits Norstedt's hand-operated belt generators and those with a 220 V motor